Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Methoxybenzylamine

Catalog No. M11103100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11103100G 100 g
M1110325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11103100G Supplier Sigma Aldrich Supplier No. M11103100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-methoxybenzylamine is a primary amine and can be used for:
  • Amination reaction of functionalized aryl bromides
  • Synthesis of functionalized organopolyphosphazenes for in vivo applications
  • Synthesis of analogs of myoseverin, 8-azapurine, for their antiproliferative activities
  • Studying the deprotection procedures of amino protecting groups such as 2-nitro- and 2,4-dinitrobenzenesulfonamides

Specifications

CAS 2393-23-9
Density 1.05 g/mL (at 25°C (literature))
Boiling Point 236°C to 237°C (lit.)
Molecular Formula C8H11NO
Linear Formula CH3OC6H4CH2NH2
Refractive Index n20/D 1.546 (literature)
Quantity 100 g
MDL Number MFCD00008122
Molecular Weight (g/mol) 137.18
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.