Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenol, 99%, Spectrum™ Chemical
SDP

Catalog No. 18608264 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
25 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-264 25 kg Amber Glass Bottle
18-608-263 100 g Amber Glass Bottle
18-608-265 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-608-264 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M228025KGBL
Add to Cart
Add to Cart

Chemical Identifiers

CAS 150-76-5

Specifications

CAS Max % 1
Melting Point 54° to 57°C
Boiling Point 243 to 246°C
Packaging Amber Glass Bottle
Quantity 25 kg
Percent Purity 0.99
Grade Reagent
Physical Form Crystalline Powder or Lumps

CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.