Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenyl beta-D-Galactopyranoside 98.0+%, TCI America™
SDP

Catalog No. M148125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M148125G 25 g
M14815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M148125G Supplier TCI America Supplier No. M148125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3150-20-7
Molecular Formula C13H18O7
Molecular Weight (g/mol) 286.28
MDL Number MFCD06797126
InChI Key SIXFVXJMCGPTRB-KSSYENDESA-N
PubChem CID 11300643
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 160°C
Color White
Quantity 25 g
Formula Weight 286.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl beta-D-Galactopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.