Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Methoxyphenyl)thiourea 98.0+%, TCI America™
SDP

Catalog No. M259325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M259325G 25 g
M25935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M259325G Supplier TCI America Supplier No. M259325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2293-07-4
Molecular Formula C8H10N2OS
Molecular Weight (g/mol) 182.24
MDL Number MFCD00004936
InChI Key SRYLJBWDZZMDSK-UHFFFAOYSA-N
Synonym 1-4-methoxyphenyl thiourea, 4-methoxyphenyl thiourea, n-4-methoxyphenyl thiourea, 1-4-methoxyphenyl-2-thiourea, p-anisylthiourea, n-p-anisylthiourea, p-methoxyphenyl thiourea, thiourea, 4-methoxyphenyl, n-p-methoxyphenyl thiourea
PubChem CID 667549
IUPAC Name (4-methoxyphenyl)thiourea
SMILES COC1=CC=C(C=C1)NC(=S)N

Specifications

Melting Point 208°C
Quantity 25 g
UN Number 2811
Formula Weight 182.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (4-Methoxyphenyl)thiourea
RTECSNumber YT6825000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.