Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M125225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1252-25G 25 g
M12525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1252-25G Supplier TCI America Supplier No. M125225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5720-07-0
Molecular Formula C7H9BO3
MDL Number MFCD00039139
InChI Key VOAAEKKFGLPLLU-UHFFFAOYSA-N
PubChem CID 201262
IUPAC Name (4-methoxyphenyl)boronic acid

Specifications

Quantity 25 g
Synonym 4-methoxyphenyl boronic acid, 4-methoxybenzeneboronic acid, p-anisylboronic acid, p-methoxyphenylboronic acid, p-methoxybenzeneboronic acid
SMILES B(C1=CC=C(C=C1)OC)(O)O
Molecular Weight (g/mol) 151.956
Formula Weight 151.96
Physical Form Crystalline Powder
RTECSNumber CY8975000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.