Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenylphosphonic Dichloride 90.0+%, TCI America™
SDP

Catalog No. M11561G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11561G 1 g
M11565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11561G Supplier TCI America Supplier No. M11561G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37632-18-1
Molecular Formula C7H7Cl2O2P
Molecular Weight (g/mol) 225.00
MDL Number MFCD00123296
InChI Key LOFIFCKKPQFWSH-UHFFFAOYSA-N
Synonym 4-Dichlorophosphonylanisole
PubChem CID 216711
IUPAC Name (4-methoxyphenyl)phosphonoyl dichloride
SMILES COC1=CC=C(C=C1)P(Cl)(Cl)=O

Specifications

Quantity 1 g
UN Number 3265
Formula Weight 225.00
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenylphosphonic Dichloride
RTECSNumber TA1825000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.