Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxythioanisole 98.0+%, TCI America™
SDP

Catalog No. M185225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M185225G 25 g
M18525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M185225G Supplier TCI America Supplier No. M185225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1879-16-9
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.227
MDL Number MFCD00010587
InChI Key DQNSKXYRRRCKGH-UHFFFAOYSA-N
Synonym 4-methoxythioanisole, 1-methoxy-4-methylthio benzene, 4-methylthio anisole, p-methylthio anisole, p-anisyl methyl sulfide, p-methoxyphenyl methyl sulfide, 1-methoxy-4-methylthiobenzene, benzene, 1-methoxy-4-methylthio, anisole, p-methylthio, 4-methoxy thioanisole
PubChem CID 15885
IUPAC Name 1-methoxy-4-methylsulfanylbenzene
SMILES COC1=CC=C(C=C1)SC

Specifications

Color Yellow
Boiling Point 137°C
Quantity 25 g
Formula Weight 154.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methoxythioanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.