Learn More
4-Methoxythiophenol, 97%
Description
4-Methoxythiophenol is used in preparing samples for self-assembled monolayers (SAM?s) characterization and to study SAM’s effect on both n- and p-channel organic thin film transistors. It is also used in cesium fluoride-Celite catalyzed preparation of thioethers and thioesters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Methoxythiophenol is used in preparing samples for self-assembled monolayers (SAM′s) characterization and to study SAM’s effect on both n- and p-channel organic thin film transistors. It is also used in cesium fluoride-Celite catalyzed preparation of thioethers and thioesters.
Solubility
Not miscible or difficult to mix.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
Specifications
Specifications
| Density | 1.14 |
| Boiling Point | 100°C to 103°C (13 mmHg) |
| Flash Point | 96°C (204°F) |
| Odor | Unpleasant |
| Refractive Index | 1.5831 |
| Quantity | 25 g |
| Beilstein | 1446910 |
| Sensitivity | Air Sensitive |
| Solubility Information | Not miscible or difficult to mix. |
| Formula Weight | 140.21 |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: III |
| EINECSNumber | 211-799-2 |
| RTECSNumber | DC1800000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.