Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-1,2,3-thiadiazole-5-carbonyl chloride, Thermo Scientific™

Catalog No. CC02302DE
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC02302DE 5 g Amber Glass Bottle
CC02302DA 1 g Amber Glass Bottle
CC02302EB 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC02302DE Supplier Thermo Scientific Chemicals Supplier No. CC02302DE
Add to Cart

Chemical Identifiers

CAS 59944-65-9
Molecular Formula C4H3ClN2OS
Molecular Weight (g/mol) 162.59
MDL Number MFCD00173831
InChI Key SDNDOCTUXWLDIX-UHFFFAOYSA-N
Synonym 4-methyl-1,2,3-thiadiazole-5-carbonyl chloride, 4-methyl-1,2,3-thiadiazole-5-carbonylchloride, 1,2,3-thiadiazole-5-carbonylchloride, 4-methyl, 4-methyl-1,2,3 thiadiazole-5-carbonyl chloride, 4-methyl-1,2,3-thiadiazole-5-carboxylic acid chloride, 4-methyl-5-thiadiazolecarbonyl chloride, 5-chlorocarbonyl-4-methyl-1,2,3-thiadiazole, 1,2,3-thiadiazole-5-carbonyl chloride, 4-methyl-9ci
PubChem CID 2736982
SMILES CC1=C(SN=N1)C(Cl)=O

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 5 g
IUPAC Name 4-methyl-1,2,3-thiadiazole-5-carbonyl chloride
Formula Weight 162.6
Percent Purity 90%
Chemical Name or Material 4-Methyl-1, 2, 3-thiadiazole-5-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.