Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-1-acetonaphthone 97.0+%, TCI America™
SDP

Catalog No. M120225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M120225G 25 g
M12025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M120225G Supplier TCI America Supplier No. M120225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28418-86-2
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00671556
InChI Key BKGIZYOJHJKFJP-UHFFFAOYSA-N
Synonym 1-Acetyl-4-methylnaphthalene
PubChem CID 97645
IUPAC Name 1-(4-methylnaphthalen-1-yl)ethanone
SMILES CC1=CC=C(C2=CC=CC=C12)C(=O)C

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 136°C
Quantity 25 g
Formula Weight 184.24
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Methyl-1-acetonaphthone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.