Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 97.0+%, TCI America™
SDP

Catalog No. M26471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26471G 1 g
M26475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26471G Supplier TCI America Supplier No. M26471G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 635305-48-5
Molecular Formula C11H17BO2S
Molecular Weight (g/mol) 224.13
MDL Number MFCD11045430
InChI Key FNRMPXJVCSMVMA-UHFFFAOYSA-N
Synonym 4,4,5,5-Tetramethyl-2-(4-methyl-2-thienyl)-1,3,2-dioxaborolane, 4-Methyl-2-thiopheneboronic Acid Pinacol Ester
PubChem CID 10443592
IUPAC Name 4,4,5,5-tetramethyl-2-(4-methylthiophen-2-yl)-1,3,2-dioxaborolane
SMILES CC1=CSC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 31°C
Color White-Yellow
Boiling Point 93°C
Quantity 1 g
Formula Weight 224.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.