Alfa Aesar™ 4-Methyl-2-pentanone, 99%

Manufacturer: Alfa Aesar™ A11618AP

Quantity 500mL

 View more versions of this product

Catalog No. AAA11618AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 108-10-1
    Molecular Formula C6H12O
    Formula Weight 100.16
    MDL Number MFCD00008938
    UN Number UN1245
    Beilstein 605399
    Merck Index 14,5207
    Synonym Hexone; Isobutyl methyl ketone
    Chemical Name or Material 4-Methyl-2-pentanone

    Specifications

    Formula Weight 100.16
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C6H12O
    Boiling Point 117° to 118°C
    CAS 108-10-1
    UN Number UN1245
    Beilstein 605399
    Flash Point 13°C (55°F)
    Health Hazard 2 GHS H Statement
    H225-H332-H319-H335
    Highly flammable liquid and vapor.
    Harmful if inhaled.
    Causes serious eye irritation.
    May cause respiratory irritation.
    MDL Number MFCD00008938
    Merck Index 14,5207
    Synonym Hexone; Isobutyl methyl ketone
    Melting Point −84°C
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL ISOBUTYL KETONE
    RTECS Number SA9275000
    Quantity 500mL
    TSCA TSCA
    Odor Mild
    Health Hazard 1 Danger
    Chemical Name or Material 4-Methyl-2-pentanone
    Assay 99%

    Description

    Applications
    4-methyl-2-pentanone is used as a solvent for nitrocellulose, gums, resins, varnishes, paints, lacquers, nitrocellulose, certain polymers and as a denaturing agent. It acts as a precursor for the preparation of N-(1,3-dimethylbutyl)-N ft.-phenyl-p-phenylene diamine (6PPD), which finds application as an antiozonant in tires. Its aqueous solution serves as a developer for PMMA electron beam lithography resist.

    Solubility
    Miscible with ethanol, ether, acetone, benzene and chloroform. Slightly miscible with water.

    Notes
    Incompatible with oxidizing agents and strong bases. Avoid conditions like flames and sparks.

    GHS H Statement
    H225-H332-H319-H335
    Highly flammable liquid and vapor.
    Harmful if inhaled.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit