Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-pentanone, ACS, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18608273 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
PVC
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-273 4 L PVC
18-608-276 100 mL Amber Glass Bottle
18-608-277 500 mL Amber Glass Bottle
18-608-271 1 L Amber Glass Bottle
18-608-272 4 L Amber Glass Bottle
18-608-275 20 L Amber Glass Bottle
18-608-274 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-608-273 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M12754LTPV
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(C)=O

Specifications

CAS Max % 1
Boiling Point 114°C to 118°C (237.2°F to 244.4°F)
Packaging PVC
Quantity 4 L
Residue after Evaporation 5e-05
Percent Purity 0.985
Grade ACS
Titratable Acid 0.002 meq/g
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentanone
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.