Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-pentanone, ACS Reagent, ≥98.5%, Solstice

Catalog No. 60046617 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-617 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-046-617 Supplier Solstice Supplier No. 360511500ML

missing translation for 'validMainframeMsg'

ACS Reagent,≥98.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00008938
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C

Specifications

Melting Point -80°C
Density 0.801g/mL at 25°C
Boiling Point 118°C
Flash Point 14°C (57.2°F)
Infrared Spectrum Conforms to structure
Packaging Glass Bottle
Refractive Index n20/D 1.395
Linear Formula (CH3)2CHCH2COCH3
Quantity 500 mL
UN Number UN1245
Beilstein 605399
Formula Weight 100.16g/mol
Percent Purity ≥98.5%
Chemical Name or Material 4-Methyl-2-pentanone
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338
DOTInformation class 3 - PG 2 - Methyl isobutyl ketone
RTECSNumber 203-550-1
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.