Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-phenyl-1,3-thiazole-5-sulfonyl chloride, Technical, Thermo Scientific™

Catalog No. CC29203CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC29203CB 250 mg Amber Glass Bottle
CC29203DA 1 g Amber Glass Bottle
CC29203DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC29203CB Supplier Thermo Scientific Chemicals Supplier No. CC29203CB
Add to Cart

Chemical Identifiers

CAS 690632-88-3
Molecular Formula C10H8ClNO2S2
Molecular Weight (g/mol) 273.749
InChI Key NGDQQLAVJWUYSF-UHFFFAOYSA-N
Synonym 4-methyl-2-phenylthiazole-5-sulfonyl chloride, 4-methyl-2-phenyl-thiazole-5-sulfonyl chloride, 5-thiazolesulfonyl chloride, 4-methyl-2-phenyl, 4-methyl-2-phenyl-1,3-thiazole-5-sulfonylchloride
PubChem CID 2795490
IUPAC Name 4-methyl-2-phenyl-1,3-thiazole-5-sulfonyl chloride
SMILES CC1=C(SC(=N1)C2=CC=CC=C2)S(=O)(=O)Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 273.76
Percent Purity 95%
Chemical Name or Material 4-Methyl-2-phenyl-1, 3-thiazole-5-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.