Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-phenylpyridine 97.0+%, TCI America™
SDP

Catalog No. M29521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29521G 1 g
M2952200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29521G Supplier TCI America Supplier No. M29521G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3475-21-6
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00087695
InChI Key WWMRJCUZPJJWBC-UHFFFAOYSA-N
Synonym 2-Phenyl-4-picoline
PubChem CID 77026
IUPAC Name 4-methyl-2-phenylpyridine
SMILES CC1=CC=NC(=C1)C1=CC=CC=C1

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 115°C
Quantity 1 g
Formula Weight 169.23
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Methyl-2-phenylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.