Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-3-nitrobenzyl Alcohol 96.0+%, TCI America™
SDP

Catalog No. m29165g Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29165G 5 g
M291625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29165G Supplier TCI America Supplier No. M29165G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40870-59-5
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00007177
InChI Key URCWIFSXDARYLY-UHFFFAOYSA-N
PubChem CID 123497
IUPAC Name (4-methyl-3-nitrophenyl)methanol
SMILES CC1=C(C=C(C=C1)CO)[N+](=O)[O-]

Specifications

Melting Point 43°C
Color Yellow
Boiling Point 195°C
Quantity 5 g
Formula Weight 167.16
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-3-nitrobenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.