Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-4-nitrovaleraldehyde 90.0+%, TCI America™
SDP

Catalog No. M182525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M182525G 25 g
M18255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M182525G Supplier TCI America Supplier No. M182525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57620-49-2
Molecular Formula C6H11NO3
Molecular Weight (g/mol) 145.16
MDL Number MFCD09030771
InChI Key YDVAFDALOGXWTO-UHFFFAOYSA-N
Synonym 4-Methyl-4-nitropentanal
PubChem CID 12264009
IUPAC Name 4-methyl-4-nitropentanal
SMILES CC(C)(CCC=O)[N+]([O-])=O

Specifications

Boiling Point 90°C
Quantity 25 g
Formula Weight 145.16
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-4-nitrovaleraldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.