Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-L-phenylalanine 98.0+%, TCI America™
SDP

Catalog No. M27431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27431G 1 g
M27435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27431G Supplier TCI America Supplier No. M27431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1991-87-3
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00038575
InChI Key DQLHSFUMICQIMB-VIFPVBQESA-N
Synonym 4-methyl-l-phenylalanine, l-4-methylphenylalanine, 4-methylphenylalanine, h-phe 4-me-oh, 2s-2-amino-3-4-methylphenyl propanoic acid, phenylalanine, 4-methyl, s-2-amino-3-p-tolyl propanoic acid, l-4-me-phe-oh, l-4-methylphe, 4-methyl-l-phe-oh
PubChem CID 151513
IUPAC Name (2S)-2-amino-3-(4-methylphenyl)propanoic acid
SMILES CC1=CC=C(C[C@H](N)C(O)=O)C=C1

Specifications

Melting Point 271°C
Color White-Yellow
Quantity 1 g
Formula Weight 179.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-L-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.