Learn More
4-Methylbenzyl bromide, 98%
Description
4-Methylbenzyl bromide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Methylbenzyl bromide is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
Specifications
Specifications
| Melting Point | 33°C to 36°C |
| Density | 1.324 |
| Boiling Point | 218°C to 220°C |
| Flash Point | 97°C (206°F) |
| Quantity | 25 g |
| UN Number | UN3417 |
| Beilstein | 507389 |
| Merck Index | 14,10090 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 185.07 |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332-H314 |
| Hazard Statement | GHS H Statement H330-H314-H318 Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLYL BROMIDE, SOLID |
| EINECSNumber | 203-240-6 |
| RTECSNumber | CY9039300 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.