Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylbenzyl cyanide, 98%

Catalog No. AC126441000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126441000 100 g Glass bottle
AC126440050 5 g Glass bottle
AC126440250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC126441000 Supplier Thermo Scientific Chemicals Supplier No. 126441000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2947-61-7 | C9H9N | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2947-61-7
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00001922
InChI Key RNHKXHKUKJXLAU-UHFFFAOYSA-N
Synonym 4-methylbenzyl cyanide, 4-methylphenylacetonitrile, p-tolylacetonitrile, p-methylbenzyl cyanide, 2-4-methylphenyl acetonitrile, benzeneacetonitrile, 4-methyl, 4-methylphenyl acetonitrile, 4-methylbenzeneacetonitrile, 4-methylbenzylcyanide, 2-p-tolylacetonitrile
PubChem CID 76280
IUPAC Name 2-(4-methylphenyl)acetonitrile
SMILES CC1=CC=C(CC#N)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 18°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 242°C to 243°C
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.5170 to 1.5195
Linear Formula CH3C6H4CH2CN
Quantity 100 g
Beilstein 09, 530
Specific Gravity 0.99
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether,benzene
Formula Weight 131.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methylbenzyl cyanide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 220-963-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.