Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylbenzyl Phenyl Ketone 97.0+%, TCI America™
SDP

Catalog No. M163925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M163925G 25 g
M16395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M163925G Supplier TCI America Supplier No. M163925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2430-99-1
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
InChI Key WILFDKCJCDVGQX-UHFFFAOYSA-N
Synonym 2-(p-Tolyl)acetophenone, p-Tolylmethyl Phenyl Ketone
PubChem CID 242473
IUPAC Name 2-(4-methylphenyl)-1-phenylethanone
SMILES CC1=CC=C(C=C1)CC(=O)C2=CC=CC=C2

Specifications

Melting Point 95°C
Color White
Boiling Point 155°C
Quantity 25 g
Formula Weight 210.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylbenzyl Phenyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.