Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylcinnamic acid, predominantly trans, 99%

Catalog No. AAA1333714
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1333714 25 g
AAA1333706 5 g
AAA1333722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1333714 Supplier Thermo Scientific Chemicals Supplier No. A1333714
Add to Cart

CAS: 1866-39-3 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1866-39-3
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00002697
InChI Key RURHILYUWQEGOS-VOTSOKGWSA-N
Synonym 4-methylcinnamic acid, 3-p-tolyl acrylic acid, e-3-p-tolyl acrylic acid, p-methylcinnamic acid, p-methyl cinnamic acid, 2e-3-4-methylphenyl acrylic acid, 2e-3-4-methylphenyl prop-2-enoic acid, 4-methyl-cinnamic acid, e-3-4-methylphenyl-2-propenoic acid, 3-4-methylphenyl-2-propenoic acid
PubChem CID 731767
IUPAC Name (E)-3-(4-methylphenyl)prop-2-enoic acid
SMILES CC1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 196°C to 200°C
Quantity 25 g
Beilstein 1364327
Formula Weight 162.19
Percent Purity 99%
Chemical Name or Material 4-Methylcinnamic acid, predominantly trans
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-479-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.