Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylsulfinyl)benzeneboronic acid, 98%

Catalog No. AAL1786503
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1786503 1 g
AAL17865MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1786503 Supplier Thermo Scientific Chemicals Supplier No. L1786503
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 166386-48-7 | C7H9BO3S | 184.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166386-48-7
Molecular Formula C7H9BO3S
Molecular Weight (g/mol) 184.02
MDL Number MFCD02093071
InChI Key YOTGALZTDVXUKZ-UHFFFAOYNA-N
Synonym 4-methanesulfinyl benzeneboronic acid, 4-methylsulfinyl phenyl boronic acid, 4-methylsulphinyl benzeneboronic acid, 4-methylsulfinylphenyl boronic acid, 4-methanesulphinylphenylboronic acid, 4-methylsulfinyl phenylboronic acid, boronic acid, 4-methylsulfinyl phenyl, 4-boronophenyl methyl sulfoxide, 4-methanesulfinylphenylboronic acid
PubChem CID 2773531
IUPAC Name (4-methylsulfinylphenyl)boronic acid
SMILES CS(=O)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point ∼170°C
Quantity 1 g
Formula Weight 184.02
Percent Purity 98%
Chemical Name or Material 4-(Methylsulfinyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.