Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylsulfonyl-2-nitrotoluene, 99%

Catalog No. AAB2144418
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2144418 50 g
AAB2144409 10 g
AAB2144430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2144418 Supplier Thermo Scientific Chemicals Supplier No. B2144418
Add to Cart

CAS: 1671-49-4 | C8H9NO4S | 215.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1671-49-4
Molecular Formula C8H9NO4S
Molecular Weight (g/mol) 215.22
MDL Number MFCD00047808
InChI Key OXBDLEXAVKAJFD-UHFFFAOYSA-N
Synonym 4-methylsulfonyl-2-nitrotoluene, 1-methyl-4-methylsulfonyl-2-nitrobenzene, 2-nitro-4-methylsulfonyltoluene, 4-mesyl-2-nitrotoluene, 4-methanesulfonyl-1-methyl-2-nitrobenzene, methyl 3-nitro-p-tolyl sulfone, benzene, 1-methyl-4-methylsulfonyl-2-nitro, 2-nitro-4-methylsulf, ksc532k5r, 4-methy-2-nitrosulfonyl toluene
PubChem CID 687125
SMILES CC1=CC=C(C=C1[N+]([O-])=O)S(C)(=O)=O

Specifications

Melting Point 119°C to 122°C
Quantity 50 g
IUPAC Name 4-methanesulfonyl-1-methyl-2-nitrobenzene
Formula Weight 215.22
Percent Purity 99%
Chemical Name or Material 4-Methylsulfonyl-2-nitrotoluene
Hazard Category H302-H334-H361f
Hazard Statement GHS H Statement
H361-H302-H317
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P281-P285-P301+P312-P304+P341-P308+P313-P330-P342+P311-P501c
EINECSNumber 430-550-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.