Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylsulfonyl)aniline 98.0+%, TCI America™
SDP

Catalog No. A2725100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2725100G 100 g
A272525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2725100G Supplier TCI America Supplier No. A2725100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5470-49-5
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00025353
InChI Key XJEVFFNOMKXBLU-UHFFFAOYSA-N
Synonym 4-methylsulfonyl aniline, 4-methylsulphonyl aniline, 4-mesylaniline, 4-methanesulfonylaniline, p-methylsulfonyl aniline, benzeneamine, 4-methylsulfonyl, benzenamine, 4-methylsulfonyl, 4-methylsulfonyl benzenamine, 4-methylsulphonylaniline
PubChem CID 79617
IUPAC Name 4-methanesulfonylaniline
SMILES CS(=O)(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 137°C
Color White
Quantity 100 g
Formula Weight 171.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylsulfonyl)aniline
EINECSNumber (3)-2137
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.