Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylsulfonyl)phenol, 95%, Thermo Scientific™

Catalog No. AC424880025
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424880025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC424880025 Supplier Thermo Scientific Chemicals Supplier No. 424880025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14763-60-1
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00047806
InChI Key KECCFSZFXLAGJS-UHFFFAOYSA-N
Synonym 4-methylsulfonyl phenol, 4-methanesulfonylphenol, 4-methanesulfonyl-phenol, 4-hydroxyphenyl methyl sulfone, 4-methylsulphonylphenol, phenol, p-methylsulfonyl, phenol, 4-methylsulfonyl, p-hydroxyphenyl methyl sulfone, p-methylsulfonyl phenol, unii-f7bif7zqj9
PubChem CID 123350
IUPAC Name 4-methylsulfonylphenol
SMILES CS(=O)(=O)C1=CC=C(O)C=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 90.0°C to 95.0°C
Color Brown-Pink to Tan
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H4SO2CH3
Quantity 2.5 g
Solubility Information Solubility in water: slight. Other solubilities: soluble in dichloromethane,methanol
Formula Weight 172.2
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-(Methylsulfonyl)phenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.