Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylsulfonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M19725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1972-5G 5 g
M19721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1972-5G Supplier TCI America Supplier No. M19725G
Add to Cart
Add to Cart

Specifications

CAS 149104-88-1
Melting Point 293°C
Color White-Yellow
Molecular Formula C7H9BO4S
MDL Number MFCD01630820
Quantity 5 g
Synonym 4-methylsulfonyl phenylboronic acid, 4-methanesulfonyl phenylboronic acid, 4-methylsulfonyl phenyl boronic acid, 4-methanesulfonyl benzeneboronic acid, 4-methanesulfonylphenylboronic acid
InChI Key VDUKDQTYMWUSAC-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)S(=O)(=O)C)(O)O
IUPAC Name (4-methylsulfonylphenyl)boronic acid
Molecular Weight (g/mol) 200.015
PubChem CID 2734364
Formula Weight 200.02
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.