Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylsulphonyl benzaldehyde, 95%

Catalog No. AC424900025
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424900025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC424900025 Supplier Thermo Scientific Chemicals Supplier No. 424900025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5398-77-6 | C8H8O3S | 184.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5398-77-6
Molecular Formula C8H8O3S
Molecular Weight (g/mol) 184.22
MDL Number MFCD00216485
InChI Key PSVPUHBSBYJSMQ-UHFFFAOYSA-N
Synonym 4-methylsulfonyl benzaldehyde, 4-methylsulphonyl benzaldehyde, 4-methanesulfonylbenzaldehyde, 4-methanesulfonyl-benzaldehyde, 4-methylsulfonyl-benzaldehyde, 4-methylsulphonylbenzaldehyde, p-methylsulphonyl benzaldehyde, benzaldehyde, 4-methylsulfonyl, pubchem8151
PubChem CID 220376
IUPAC Name 4-methylsulfonylbenzaldehyde
SMILES CS(=O)(=O)C1=CC=C(C=C1)C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 155°C to 161°C
Color Beige-Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 2.5 g
Solubility Information Solubility in water: insoluble
Formula Weight 184.22
Percent Purity 95%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Methylsulphonyl benzaldehyde, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.