Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylsulphonyl benzonitrile, 95%, Thermo Scientific™

Catalog No. AC424890025
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424890025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC424890025 Supplier Thermo Scientific Chemicals Supplier No. 424890025
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22821-76-7
Molecular Formula C8H7NO2S
Molecular Weight (g/mol) 181.21
MDL Number MFCD00216489
InChI Key FARXIDYHJAANGP-UHFFFAOYSA-N
Synonym 4-methylsulfonyl benzonitrile, 4-methanesulfonyl-benzonitrile, 4-methanesulfonylbenzonitrile, benzonitrile, 4-methylsulfonyl, benzonitrile, p-methylsulfonyl, 4-methylsulfonyl benzenecarbonitrile, 4-mesylbenzonitrile, maybridge3_003168, p-methylsulfonylbenzonitrile, 4-methylsulphonylbenzonitrile
PubChem CID 519981
IUPAC Name 4-methylsulfonylbenzonitrile
SMILES CS(=O)(=O)C1=CC=C(C=C1)C#N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 137.0°C to 142.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 2.5 g
Solubility Information Solubility in water: negligible
Formula Weight 181.21
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylsulphonyl benzonitrile, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with p

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.