Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylthiazole, 99%

Catalog No. AAA1542814
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1542814 25 g
AAA1542806 5 g
AAA1542822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1542814 Supplier Thermo Scientific Chemicals Supplier No. A1542814
Add to Cart

CAS: 693-95-8 | C4H5NS | 99.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-95-8
Molecular Formula C4H5NS
Molecular Weight (g/mol) 99.15
MDL Number MFCD00005340
InChI Key QMHIMXFNBOYPND-UHFFFAOYSA-N
Synonym 4-methylthiazole, thiazole, 4-methyl, 4-methyl thiazole, 4-methyl-thiazole, fema no. 3716, 4-methyl-5h-1,3-thiazole, 4-methyithiazole, methylthiazole, 4, pubchem23683
PubChem CID 12748
ChEBI CHEBI:35626
IUPAC Name 4-methyl-1,3-thiazole
SMILES CC1=CSC=N1

Specifications

Density 1.093
Boiling Point 133°C to 134°C
Flash Point 32°C (89°F)
Odor Characteristic
Refractive Index 1.524
Quantity 25 g
UN Number UN1993
Beilstein 105228
Formula Weight 99.16
Percent Purity 99%
Chemical Name or Material 4-Methylthiazole
Show More Show Less
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 211-764-1
RTECSNumber XJ5096000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.