Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylthiazole 98.0+%, TCI America™
SDP

Catalog No. M089425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M089425G 25 g
M08945G 5 g
M0894100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M089425G Supplier TCI America Supplier No. M089425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 693-95-8
Molecular Formula C4H5NS
Molecular Weight (g/mol) 99.15
MDL Number MFCD00005340
InChI Key QMHIMXFNBOYPND-UHFFFAOYSA-N
Synonym 4-methylthiazole, thiazole, 4-methyl, 4-methyl thiazole, 4-methyl-thiazole, fema no. 3716, 4-methyl-5h-1,3-thiazole, 4-methyithiazole, methylthiazole, 4, pubchem23683
PubChem CID 12748
ChEBI CHEBI:35626
IUPAC Name 4-methyl-1,3-thiazole
SMILES CC1=CSC=N1

Specifications

Color Yellow
Boiling Point 134°C
Quantity 25 g
UN Number 1993
Formula Weight 99.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methylthiazole
RTECSNumber XJ5096000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.