Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylthio)phenol 98.0+%, TCI America™
SDP

Catalog No. M0441250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0441250G 250 g
M044125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0441250G Supplier TCI America Supplier No. M0441250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1073-72-9
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.20
MDL Number MFCD00002351
InChI Key QASBCTGZKABPKX-UHFFFAOYSA-N
Synonym 4-methylthio phenol, 4-methylmercapto phenol, phenol, 4-methylthio, p-methylthio phenol, 4-hydroxythioanisole, 4-methylmercaptophenol, p-hydroxythioanisole, 4-methylsulfanyl phenol, phenol, p-methylthio, p-hydroxyphenyl methyl sulfide
PubChem CID 14086
ChEBI CHEBI:38862
IUPAC Name 4-(methylsulfanyl)phenol
SMILES CSC1=CC=C(O)C=C1

Specifications

Melting Point 86°C
Color White-Yellow
Boiling Point 154°C
Quantity 250 g
Formula Weight 140.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylthio)phenol
EINECSNumber (3)-1099
RTECSNumber SM1578500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.