Learn More
Sigma Organic Chemistry 4-methylthioph -2-boron | 250mg | 162607-15-0 | MFCD01319040
Supplier: Sigma Organic Chemistry CDS003645250MG

4-methylthioph -2-boron , 250mg
About this Item:
CAS #: 162607-15-0
MDL #: MFCD01319040
Molecular Weight: 141.98
UNSPSC Code: 12352200
Chemical Formula: C5H7BO2S
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.