Learn More
Medchemexpress LLC 4-Methylumbelliferyl 100mg | 18323-58-5 | 440.61 g·mol^-1 | C28H40O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY117095100MG
Encompass_Preferred
4-Methylumbelliferyl oleate is a fluorogenic substrate for acid and alkaline lipases. Upon enzymatic hydrolysis, it liberates 4-methylumbelliferone (4-MU), enabling sensitive fluorometric detection and kinetic assays (Ex/Em 320/450 nm).
  • Provides a fluorogenic readout upon lipase-catalyzed hydrolysis.
  • Releases 4-methylumbelliferone with Ex/Em 320/450 nm for sensitive detection.
  • High chemical purity suitable for enzymatic assays.
  • Stable when stored at -20°C and protected from light; solvent stability guidelines available.
  • Available in multiple pack sizes to support assay development and scale-up.
Catalog No. 50-003-08390
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.