Learn More
Abcam 4-Methylumbelliferyl 2-Acetamido-2-deoxy-alpha-D-glucopyranoside, Fluorogenic Substrate, 10 mg
Description
- Molecular Weight: 379.36 g/mol
- Applications: Fluorogenic substrate for assays of N-Acetyl-α-D-glucosaminidase
- Research Use: Can be used to test for Sanfilippo B Syndrome
- Composition: 100% 4-Methylumbelliferyl 2-Acetamido-2-deoxy-alpha-D-Glucopyranoside
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | N-Acetyl-α-D-glucosaminidase |
| Substrate Properties | Chemical Substrate |
| Substrate | 4-Methylumbelliferyl 2-acetamido-2-deoxy-alpha-D-glucopyranoside |
| Buffer | 100% 4-Methylumbelliferyl 2-Acetamido-2-deoxy-alpha-D-Glucopyranoside |
| Content And Storage | Store at -20°C. |
| Shipping Condition | Ambient |
| Detection Method | Fluorescence |
| Formulation | 100% 4-Methylumbelliferyl 2-Acetamido-2-deoxy-alpha-D-Glucopyranoside |
| Quantity | 10 mg |
| For Use With (Application) | N-Acetyl-α-D-glucosaminidase assays, Sanfilippo B Syndrome testing |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.