Learn More
Medchemexpress LLC 4-Methylumbelliferyl | 124223-99-0 | MFCD00270047 | 99.0% | 379.36 g/mol | C18H21NO8 | 5MG
Supplier: Medchemexpress LLC HY1230005MG
4-Methylumbelliferyl 2-acetamido-2-deoxy-α-D-galactopyranoside is a fluorogenic glycoside substrate that liberates fluorescent 4-methylumbelliferone upon α-galactosidase-mediated hydrolysis, enabling sensitive fluorometric measurement of enzyme activity in biochemical and cell-based assays.
- Fluorogenic substrate for α-galactosidase assays.
- Releases 4-methylumbelliferone for sensitive fluorescence detection.
- High purity suitable for quantitative measurements.
- Supplied in small-quantity packaging suitable for assay development.
- Solid form; handle and store according to standard glycoside reagent guidelines.