Learn More
Sigma Aldrich Fine Chemicals Biosciences 4-Methylumbelliferyl alpha-D-glucopyranoside alpha-glucosidase substrate | 17833-43-1 | MFCD00067661 | 1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M97661G
Encompass
4-Methylumbelliferyl alpha-D-glucopyranoside alpha-glucosidase substrate | Purity: >=99% (TLC) | Mol Wt: 338.31 | 17833-43-1 | MFCD00067661 | 1G
Catalog No. 50-178-9560