Learn More
Cayman Chemical 4-Methylumbelliferyl-β-D-Glucuronide (hydrate), 250 mg
SDP
Supplier:  Cayman Chemical 17203250
Encompass_Preferred
A fluorogenic substrate of β-glucuronidase; cleaved by β-glucuronide to release the fluorescent moiety 4-MU
Catalog No. NC1288394
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.