Learn More
Abcam 4-Methylumbelliferyl Heptanoate, Fluorogenic Lipase Substrate, 100 mg
Description
- Molecular weight: 288.34 g/mol
- Purity: >95%
- Application: Fluorescent substrate for esterases and lipases; also used for measuring cell proliferation and viability
- Composition: 100% 4-Methylumbelliferyl heptanoate
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Esterase, Lipase |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Substrate | 4-Methylumbelliferyl heptanoate |
| Content And Storage | Store at -20°C. |
| Detection Method | Fluorescence |
| Buffer | 100% 4-Methylumbelliferyl heptanoate |
| Formulation | 100% 4-Methylumbelliferyl heptanoate |
| Quantity | 100 mg |
| For Use With (Application) | Esterase activity measurement, Lipase activity measurement, Cell proliferation measurement, Cell viability measurement |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.