Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 4-Methylumbelliferyl Heptanoate, Fluorogenic Lipase Substrate, >95% Purity, 500 mg

Catalog No. 502705162
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5162 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5162 Supplier Abcam Supplier No. AB145482500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Fluorescent substrate for esterases and lipases used to measure cell proliferation and viability.

  • Molecular weight: 288.34 g/mol
  • Purity: >95%
  • Application: Fluorescent substrate for esterases and lipases; also used to measure cell proliferation and viability
  • Storage buffer: 100% 4-Methylumbelliferyl heptanoate

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Esterases, Lipases
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Substrate 4-Methylumbelliferyl heptanoate
Buffer 100% 4-Methylumbelliferyl heptanoate
Form Powder
Detection Method Fluorescence
Content And Storage Store at -20°C.
Formulation 100% 4-Methylumbelliferyl heptanoate
Quantity 500 mg
For Use With (Application) Esterase and lipase assay, Cell proliferation measurement, Cell viability measurement
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.