Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 4-Methylumbelliferyl Oleate, Fluorogenic Lipase Substrate, 25 mg

Catalog No. 502705242
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5242 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5242 Supplier Abcam Supplier No. AB14538525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorogenic substrate for lipases with >95% purity suitable for use in aqueous or organic solvents.

  • Molecular Weight: 440.61 g/mol
  • Purity: >95%
  • Application: Fluorogenic substrate for lipases
  • Compatibility: Suitable for use in aqueous solutions and several organic solvents
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Lipases
Shipping Condition Ambient
Substrate 4-Methylumbelliferyl oleate
Content And Storage Store at 4°C.
Detection Location In-Solution Detection
Detection Method Fluorescence
Buffer 100% 4-methylumbelliferyl oleate
Formulation 100% 4-Methylumbelliferyl oleate
Quantity 25 mg
For Use With (Application) Lipase assay
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.