Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 4-Methylumbelliferyl Oleate, Fluorogenic Lipase Substrate, 250 mg

Catalog No. 502705243
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5243 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5243 Supplier Abcam Supplier No. AB145385250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorogenic substrate for lipases that is usable in both aqueous solutions and several organic solvents.

  • Molecular Weight: 440.61 g/mol
  • Purity: >95%
  • Application: Fluorogenic substrate for lipases
  • Solubility: Suitable for use in aqueous solutions and several organic solvents

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate 4-Methylumbelliferyl oleate
Shipping Condition Ambient
Substrate Properties Lipid-based Substrate
Target Molecule Lipase
Content And Storage Store at 4°C.
Detection Method Fluorescence
Quantity 250 mg
For Use With (Application) Lipase assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.