Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Morpholinomethyl)aniline, 95+%, Thermo Scientific™

Catalog No. CC29314DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC29314DE 5 g Amber Glass Bottle
CC29314CB 250 mg Amber Glass Bottle
CC29314DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC29314DE Supplier Thermo Scientific Chemicals Supplier No. CC29314DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51013-67-3
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
InChI Key WNYFVEFUHMDIRQ-UHFFFAOYSA-N
Synonym 4-morpholinomethyl aniline, 4-morpholin-4-ylmethyl aniline, 4-morpholin-4-ylmethylphenylamine, 4-morpholin-4-ylmethyl-phenylamine, benzenamine, 4-4-morpholinylmethyl, 4-4-morpholinylmethyl aniline, 4-morpholin-4-ylmethyl-aniline, 4-morpholin-4-ylmethyl phenylamine, acmc-209krj, 4-morpholinomethyl-aniline
PubChem CID 776851
IUPAC Name 4-(morpholin-4-ylmethyl)aniline
SMILES C1COCCN1CC2=CC=C(C=C2)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 192.26
Percent Purity 95%
Chemical Name or Material 4-(Morpholinomethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.