Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Morpholinomethyl)benzoic acid, Thermo Scientific™

Catalog No. CC29301CB
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC29301CB 250 mg Amber Glass Bottle
CC29301DA 1 g Amber Glass Bottle
CC29301EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC29301CB Supplier Thermo Scientific Chemicals Supplier No. CC29301CB
Add to Cart

Chemical Identifiers

CAS 62642-62-0
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.256
InChI Key QYBXZYYECZFQRX-UHFFFAOYSA-N
Synonym 4-morpholinomethyl benzoic acid, 4-morpholin-4-ylmethyl benzoic acid, 4-morpholin-4-ylmethyl-benzoic acid, 4-4-morpholinemethyl benzoic acid, 4-4-morpholinylmethyl benzoic acid, benzoic acid, 4-4-morpholinylmethyl, timtec-bb sbb011465, cbmicro_000050, acmc-1b2v0, 4-morpholinomethylbenzoic acid
PubChem CID 703507
IUPAC Name 4-(morpholin-4-ylmethyl)benzoic acid
SMILES C1COCCN1CC2=CC=C(C=C2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 221.25
Percent Purity 97%
Chemical Name or Material 4-(Morpholinomethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.