Learn More
Sigma Organic Chemistry 4-n-Butyl thiobenzamide | 1G | MFCD09025670
Supplier: Sigma Organic Chemistry BOG000831G

4-n-Butyl thiobenzamide , 1G
About this Item:
MDL #: MFCD09025670
Molecular Weight: 193.31
UNSPSC Code: 12352200
Chemical Formula: C11H15NS
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.