Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-n-Hexadecyloxyaniline, 98%, Thermo Scientific™

Catalog No. AAL0897004
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0897004 2 g
AAL0897009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0897004 Supplier Thermo Scientific Chemicals Supplier No. L0897004

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7502-06-9
Molecular Formula C22H39NO
Molecular Weight (g/mol) 333.56
MDL Number MFCD00025325
InChI Key DETPMDIIFZGDAE-UHFFFAOYSA-N
Synonym p-hexadecyloxyaniline, 4-n-hexadecyloxyaniline, 4-hexadecyloxy aniline, 4-hexadecyloxyphenylamine, s-2-4-2,4-diaminopteridin-9-yl methyl methyl amino benzamido pentanedioic acid
PubChem CID 139038
IUPAC Name 4-hexadecoxyaniline
SMILES CCCCCCCCCCCCCCCCOC1=CC=C(C=C1)N

Specifications

Melting Point 59°C to 61°C
Quantity 2 g
Beilstein 2655373
Formula Weight 333.56
Percent Purity 98%
Chemical Name or Material 4-n-Hexadecyloxyaniline
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.