Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(N,N-Diphenylamino)benzaldehyde 98.0+%, TCI America™
SDP

Catalog No. D217225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D217225G 25 g
D21725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D217225G Supplier TCI America Supplier No. D217225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4181-05-9
Molecular Formula C19H15NO
Molecular Weight (g/mol) 273.34
MDL Number MFCD00145131
InChI Key UESSERYYFWCTBU-UHFFFAOYSA-N
Synonym 4-diphenylamino benzaldehyde, 4-n,n-diphenylamino benzaldehyde, 4-diphenylaminobenzaldehyde, 4-diphenylamino-benzaldehyde, benzaldehyde, 4-diphenylamino, 4-formyltriphenylamine, p-formyltriphenylamine, 4-n-phenylanilino benzaldehyde, p-diphenylaminobenzaldehyde, 4-n,n-diphenylamino-benzaldehyde
PubChem CID 77846
IUPAC Name 4-(N-phenylanilino)benzaldehyde
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O

Specifications

Melting Point 132°C
Quantity 25 g
Formula Weight 273.34
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(N,N-Diphenylamino)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.