Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-n-Octyloxybenzoic acid, 98%, Thermo Scientific™

Catalog No. AAB2265918
Click to view available options
Quantity:
10 g
50 g

Catalog No. AAB2265918

Supplier: Thermo Scientific Chemicals B2265918

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2493-84-7
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.34
MDL Number MFCD00013993
InChI Key IALWCYFULVHLEC-UHFFFAOYSA-N
Synonym 4-octyloxybenzoic acid, 4-n-octyloxybenzoic acid, 4-octyloxy benzoic acid, p-octyloxybenzoic acid, p-octoxybenzoic acid, p-n-octyloxybenzoic acid, p-octyloxy benzoic acid, benzoic acid, 4-octyloxy, p-n-octyloxy benzoic acid, benzoic acid, p-octyloxy
PubChem CID 17231
IUPAC Name 4-octoxybenzoic acid
SMILES CCCCCCCCOC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 101°C to 105°C
Quantity 50 g
Formula Weight 250.34
Percent Purity 98%
Chemical Name or Material 4-n-Octyloxybenzoic acid
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.