Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-n-Octyloxyphenol 98.0+%, TCI America™
SDP

Catalog No. O014025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O014025G 25 g
O01405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O014025G Supplier TCI America Supplier No. O014025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3780-50-5
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.328
MDL Number MFCD00045779
InChI Key HFRUPPHPJRZOCM-UHFFFAOYSA-N
Synonym 4-octyloxy phenol, 4-n-octyloxy phenol, 4-n-octyloxyphenol, phenol, 4-octyloxy, 4-octyloxyphenol, p-octyloxyphenol, p-octyloxy phenol, 4-n-octoxyphenol, pubchem13211, acmc-209ivj
PubChem CID 77412
IUPAC Name 4-octoxyphenol
SMILES CCCCCCCCOC1=CC=C(C=C1)O

Specifications

Melting Point 60°C
Boiling Point 127°C
Quantity 25 g
Formula Weight 222.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-n-Octyloxyphenol
EINECSNumber (3)-3539
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.